Orthophosphoric acid ≥85% (w/w) in water
Supplier: BeanTown Chemical
CAS: 7664-38-2; EC No: 231-633-2; MDL No: MFCD00011340; RTECS: TB6300000
UN No: UN1805; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: H3PO4; MW: 98.00
Melting Point: 19-21°
Density (g/mL): 1.685; Refractive Index: 1.431
- RTECS: TB6300000
- Refractive index: 1.431
- Liquid
Formula:
H₃PO₄ Melting Pt: 19…21 °C Density: 1.685 g/mL |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...