Nickel(II) carbonate basic hydrate Ni 46%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Nickel(II) carbonate hydroxide hydrate, Basic nickel carbonate

211160-1KG 211160-250G
BT211160-1KGEA 116 USD
BT211160-1KG BT211160-250G
Nickel(II) carbonate basic hydrate Ni 46%, Technical Grade
Nickel (II) carbonate basic hydrate

CAS: 12607-70-4; EC No: 235-715-9; MDL No: MFCD00016262; RTECS: QR6250000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: NiCO3·2Ni(OH)2·xH2O; MW: 304.11 (anhydrous)


  • RTECS: QR6250000
  • Powder

Formula: CH₄Ni₃O₇·H₂O
MW: 304.11 g/mol (anhy)
MDL Number: MFCD00150262
CAS Number: 39430-27-8
EINECS: 235-715-9
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR