Indium antimonide ≥99.999%

Supplier: BeanTown Chemical

145925-1G 145925-5G
BT145925-1GEA 24 USD
BT145925-1G BT145925-5G
Indium antimonide ≥99.999%
Indium antimonide

CAS: 1312-41-0; EC No: 215-192-3; MDL No: MFCD00016146 UN No: UN1549; Haz Class: 6.1; Packing Group: III 6mm & Down Pieces; Linear Formula: InSb; MW: 236.57 Melting Point: 535°


  • 6mm & Down Pieces

Formula: InSb
MW: 236.58
Melting Pt: 527
Density: 5.78
MDL Number: MFCD00016146
CAS Number: 1312-41-0
EINECS: 215-192-3
UN: 1549
ADR: 6.1 III
Merck Index: 13,04972

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR