Perchloric acid 0.1 N acetic acid Volumetric
Supplier: BeanTown Chemical
145850-1L
145850-4L
BT145850-1LEA
94.64
USD
BT145850-1L
BT145850-4L
Perchloric acid 0.1 N acetic acid Volumetric
Perchloric acid
CAS: 7601-90-3; EC No: 231-512-4; MDL No: MFCD00011325
UN No: UN2920; Haz Class: 8 (3); Packing Group: II
Liquid; Molecular Formula: HClO4 ; MW: 100.46
Boiling Point: 120°; Flash point: 40°C (104°F)
Density (g/mL): 1.05
- Liquid
Formula:
HClO₄ MW: 100.46 g/mol Boiling Pt: 39 Melting Pt: -101 Density: 1.05 g/mL Flash Pt: 40 °C (104 °F) |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 2920 ADR: 8 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...