Terbium ≥99.9% (trace rare earth metals basis), powder -40 mesh

Supplier: BeanTown Chemical

145700-1G 145700-5G
BT145700-1GEA 78.69 USD
BT145700-1G BT145700-5G
Terbium ≥99.9% (trace rare earth metals basis), powder -40 mesh
Terbium

  • Powder, packaged in air stable poly bottle

Formula: Tb
MW: 158.93 g/mol
Boiling Pt: 3230 °C
Melting Pt: 1356 °C
Density: 8.234 g/mL
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR