Palladium(II) chloride (9 - 11% Pd) (w/w) in water
Supplier: BeanTown Chemical
Synonyms:
Palladium dichloride
145465-C1G
145465-C5G
BT145465-C1GEA
168.24
USD
BT145465-C1G
BT145465-C5G
Palladium(II) chloride (9 - 11% Pd) (w/w) in water
Palladium (II) chloride
- Liquid
Formula:
PdCl₂ MW: 177.33 g/mol Density: 1.06 g/mL |
MDL Number:
MFCD00003558 CAS Number: 7647-10-1 EINECS: 231-596-2 UN: 3264 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...