1,2-Dichloroethane ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Ethylene chloride, Ethylene dichloride, Dichloroethane
144980-1L
144980-4L
144980-500ML
BT144980-1LEA
69.19
USD
BT144980-1L
BT144980-4L
BT144980-500ML
1,2-Dichloroethane ≥99.0% ACS
1,2-Dichloroethane
CAS: 107-06-2; EC No: 203-458-1; MDL No: MFCD00000963; RTECS: KI0525000
UN No: UN1184; Haz Class: 3 (6.1); Packing Group: II
Liquid; Linear Formula: ClCH2CH2Cl ; Molecular Formula: C2H4Cl2; MW: 98.96
Melting Point: -35°; Boiling Point: 83°; Flash point: 13°C (55°F)
Density (g/mL): 1.256; Refractive Index: 1.444
- RTECS: KI0525000
- Refractive index: 1.444
- Liquid
Formula:
ClH₂CCH₂Cl MW: 98.96 g/mol Boiling Pt: 83.5 Melting Pt: -35.5 Density: 1.256 g/mL Flash Pt: 13 °C (55 °F) |
MDL Number:
MFCD00000963 CAS Number: 107-06-2 EINECS: 203-458-1 UN: 1184 ADR: 3 II Merck Index: 13,03831 |
|
Specification Test Results
Assay | 99.0% min. |
Color (APHA) | ≤10 |
Residue after evaporation | ≤20ppm |
Titrable acid | ≤0.0003meq/g |
Water | ≤300ppm |
Iron (Fe) | Per lot |
Specific gravity | Per lot |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...