Cyclohexane ≥99.0% ACS
Supplier: BeanTown Chemical
CAS: 110-82-7; EC No: 203-806-2; MDL No: MFCD00003814; RTECS: GU6300000
UN No: UN1145; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6H12; MW: 84.16
Melting Point: 4-7°; Boiling Point: 80.7°; Flash point: -18°C (-1°F)
Density (g/mL): 0.779; Refractive Index: 1.426
- RTECS: GU6300000
- Refractive index: 1.426
- Liquid
Formula:
C₆H₁₂ Boiling Pt: 80.7 °C Melting Pt: 4…7 °C Density: 0.779 g/mL Flash Pt: –18 °C (–1 °F) |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II |
|
Specification Test Results
Assay | 99.0% min. |
Color (APHA) | ≤10 |
Residue after evaporation | ≤0.002% |
Substances darkened by H2SO4 | Passes test |
Water | ≤0.02% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...