Trifluoroacetic acid ≥99.5% for biochemistry

Supplier: BeanTown Chemical

Synonyms: TFA

144430-1KG 144430-250G 144430-50G
BT144430-1KGEA 384 USD
BT144430-1KG BT144430-250G BT144430-50G
Trifluoroacetic acid ≥99.5% for biochemistry
Trifluoroacetic acid

CAS: 76-05-1; EC No: 200-929-3; MDL No: MFCD00004169; RTECS: AJ9625000 UN No: UN2699; Haz Class: 8; Packing Group: I Liquid; Linear Formula: CF3COOH; MW: 114.02 Melting Point: -15°; Boiling Point: 72-73° Density (g/mL): 1.489; Refractive Index: 1.284 Hygroscopic


  • RTECS: AJ9625000
  • Refractive index: 1.284
  • Hygroscopic
  • Liquid

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8
Melting Pt: -15
Density: 1.489 g/mL
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR