4-Toluoyl chloride ≥99%

Supplier: BeanTown Chemical

Synonyms: p-Toluoyl chloride, 4-Methylbenzene-1-carbonyl chloride, p-Methylbenzoyl chloride, 4-Methylbenzoyl chloride

144400-100G 144400-2.5KG 144400-500G
BT144400-100GEA 29 USD
BT144400-100G BT144400-2.5KG BT144400-500G
4-Toluoyl chloride ≥99%
4-Toluoyl chloride

CAS: 874-60-2; EC No: 212-864-8; MDL No: MFCD00000696; RTECS: XV1660000 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Linear Formula: CH3C6H4COCl; Molecular Formula: C8H7ClO ; MW: 154.60 Melting Point: -2°; Boiling Point: 225-227°; Flash point: 82°C (179°F) Density (g/mL): 1.169; Refractive Index: 1.553 Moisture Sensitive


  • RTECS: XV1660000
  • Refractive index: 1.553
  • Moisture Sensitive
  • Liquid

Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 225
Melting Pt: -2
Density: 1.169 g/mL
Flash Pt: 82 °C (179 °F)
MDL Number: MFCD00000696
CAS Number: 874-60-2
EINECS: 212-864-8
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR