Zinc selenide ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Zinc (II) selenide

144360-10G 144360-250G 144360-50G
BT144360-10GEA 76.87 USD
BT144360-10G BT144360-250G BT144360-50G
Zinc selenide ≥99.999%
Zinc selenide

CAS: 1315-09-9; EC No: 215-259-7; MDL No: MFCD00011296 UN No: UN3283; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: ZnSe; MW: 144.35 Melting Point: 1100° Density (g/mL): 5.42 Air Sensitive, Moisture Sensitive

Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793
Density: 5.42 (g/mL)
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1 III
Merck Index: 12,10290

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR