Trichloroacetic acid (TCA) ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

144045-1KG 144045-250G 144045-50G
BT144045-1KGEA 314.67 USD
BT144045-1KG BT144045-250G BT144045-50G
Trichloroacetic acid (TCA) ≥99.0% ACS
Trichloroacetic acid (TCA)

CAS: 76-03-9; EC No: 200-927-2; MDL No: MFCD00004177; RTECS: AJ7875000 UN No: UN1839; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: CCl3COOH; MW: 163.39 Melting Point: 54-58°; Boiling Point: 196°; Flash point: 110°C (230°F) Density (g/mL): 1.629; Refractive Index: 1.62 Hygroscopic


  • RTECS: AJ7875000
  • Refractive index: 1.62
  • Hygroscopic
  • Crystalline

Formula: Cl₃CCOOH
Boiling Pt: 196 °C
Melting Pt: 54…58 °C
Density: 1.629 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR