Tetrabutylammonium tetrahydroborate ≥98%

Supplier: BeanTown Chemical

Synonyms: Tetra-n-butylammonium borohydride, Tetrabutylammonium borohydride, Tetrabutylammonium tetrahydridoborate

143595-10G 143595-250G 143595-50G
BT143595-10GEA 76 USD
BT143595-10G BT143595-250G BT143595-50G
Tetrabutylammonium tetrahydroborate ≥98%
Tetrabutylammonium tetrahydroborate

CAS: 33725-74-5; EC No: 251-658-2; MDL No: MFCD00012035 UN No: UN3131; Haz Class: 4.3 (8); Packing Group: II Crystalline; Linear Formula: [CH3(CH2)3]4N(BH4); Molecular Formula: C16H40BN; MW: 257.31 Melting Point: 124-128°; Flash point: 140°C (284°F) Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₁₆H₄₀BN
MW: 257.31 g/mol
Melting Pt: 124…128 °C
Flash Pt: 140 °C (284 °F)
MDL Number: MFCD00012035
CAS Number: 33725-74-5
EINECS: 251-658-2
UN: 3131
ADR: 4.3 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR