4-Cyano-4'-pentylbiphenyl ≥99%

Supplier: BeanTown Chemical

Synonyms: 4'-Pentyl-4-biphenylcarbonitrile, 4'-Pentyl-[1,1'-biphenyl]-4-carbonitrile

143515-1G 143515-5G
BT143515-1GEA 63 USD
BT143515-1G BT143515-5G
4-Cyano-4'-pentylbiphenyl ≥99%
4-Cyano-4'-pentylbiphenyl

CAS: 40817-08-1; EC No: 255-093-2; MDL No: MFCD00036350 UN No: UN3276; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C18H19N; MW: 249.35 Boiling Point: 140-150°/0.5 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.008; Refractive Index: 1.532 Hygroscopic


  • Refractive index: 1.532
  • Hygroscopic
  • Liquid

Formula: C₁₈H₁₉N
MW: 249.36 g/mol
Boiling Pt: 140…150 °C/0.5 mmHg
Density: 1.008 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00036350
CAS Number: 40817-08-1
EINECS: 255-093-2
ADR: 6.1 III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR