Phosphorus(III) bromide ≥99%

Supplier: BeanTown Chemical

Synonyms: Phosphorus tribromide

143500-25G 143500-100G
BT143500-100GEA 93.34 USD
BT143500-100G BT143500-25G
Phosphorus(III) bromide ≥99%
Phosphorus (III) bromide

  • RTECS: TH4460000
  • Refractive index: 1.697
  • Moisture Sensitive
  • Liquid

Formula: PBr₃
MW: 270.69 g/mol
Boiling Pt: 175 °C
Melting Pt: –41 °C
Density: 2.88 g/mL
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 1808
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR