Europium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.25 mm
Supplier: BeanTown Chemical
142890-25X25MM
142890-50X50MM
BT142890-25X25EA
362
USD
BT142890-25X25
BT142890-50X50
Europium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.25 mm
Europium
CAS: 7440-53-1; EC No: 231-161-7; MDL No: MFCD00010992
UN No: UN3208; Haz Class: 4.3; Packing Group: II
Solid; Molecular Formula: Eu; MW: 151.96
Melting Point: 822°; Boiling Point: 1527°
Density (g/mL): 5.24
- Solid
Formula:
Eu Boiling Pt: 1527 °C Melting Pt: 822 °C Density: 5.24 g/mL |
MDL Number:
MFCD00010992 CAS Number: 7440-53-1 EINECS: 231-161-7 UN: 3208 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...