m-Xylene ≥99%
Supplier: BeanTown Chemical
CAS: 108-38-3; EC No: 203-576-3; MDL No: MFCD00008536; RTECS: ZE2275000
UN No: UN1307; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: C6H4(CH3)2; Molecular Formula: C8H10; MW: 106.17
Melting Point: -48°; Boiling Point: 138-139°; Flash point: 25°C (77°F)
Density (g/mL): 0.868; Refractive Index: 1.497
- RTECS: ZE2275000
- Refractive index: 1.497
- Liquid
Formula:
C₈H₁₀ Boiling Pt: 138…139 °C Melting Pt: –48 °C Density: 0.868 g/mL Flash Pt: 25 °C (77 °F) |
MDL Number:
MFCD00008536 CAS Number: 108-38-3 EINECS: 203-576-3 UN: 1307 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...