Disulfiram ≥97%
Supplier: BeanTown Chemical
CAS: 97-77-8; EC No: 202-607-8; MDL No: MFCD00009048; RTECS: JO1225000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: (CH3CH2)2NCSS2CSN(CH2CH3)2; Molecular Formula: C10H20N2S4; MW: 296.54
Melting Point: 69-71°; Boiling Point: 117°/17 mmHg
Density (g/mL): 1.3
- RTECS: JO1225000
- Powder
Formula:
C₁₀H₂₀N₂S₄ Boiling Pt: 117 °C/17 mmHg Melting Pt: 69…71 °C Density: 1.3 g/mL |
MDL Number:
MFCD00009048 CAS Number: 97-77-8 EINECS: 202-607-8 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...