Lithium hexafluorophosphate ≥98%

Supplier: BeanTown Chemical

Synonyms: Lithium hexafluorophosphate(V)

142160-10G 142160-50G
BT142160-10GEA 59.17 USD
BT142160-10G BT142160-50G
Lithium hexafluorophosphate ≥98%
Lithium hexafluorophosphate

CAS: 21324-40-3; EC No: 244-334-7; MDL No: MFCD00011096 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II Powder; Linear Formula: LiPF6 ; MW: 151.90 Melting Point: 200° (decomposes) Density (g/mL): 1.5 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: F₆LiP
MW: 151.91 g/mol
Melting Pt: 200 °C (dec.)
Density: 1.5 g/mL
MDL Number: MFCD00011096
CAS Number: 21324-40-3
EINECS: 244-334-7
UN: 2923
ADR: 8 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR