Zinc selenide ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

142145-100G 142145-25G
BT142145-100GEA 397 USD
BT142145-100G BT142145-25G
Zinc selenide ≥99.99% (trace metals basis)
Zinc selenide

CAS: 1315-09-9; EC No: 215-259-7; MDL No: MFCD00011296 UN No: UN3283; Haz Class: 6.1; Packing Group: III -325 Mesh Powder; Linear Formula: ZnSe; MW: 144.37 Melting Point: 1102° Density (g/mL): 7.42 Air Sensitive, Moisture Sensitive

Formula: SeZn
Melting Pt: >1100°
Density: 5.42 (g/mL)
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR