Sodium dichromate dihydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium bichromate

142025-500G 142025-100G
BT142025-500GEA 113.85 USD
BT142025-500G BT142025-100G
Sodium dichromate dihydrate ≥99%
Sodium dichromate dihydrate

  • RTECS: HX7750000
  • Hygroscopic
  • Crystalline

Formula: Na₂Cr₂O₇·2H₂O
MW: 298.00 g/mol (261.97 anhy)
Melting Pt: 91 °C
Density: 2.350 g/mL
MDL Number: MFCD00149166
CAS Number: 7789-12-0
EINECS: 234-190-3
UN: 3087
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR