Iodine monochloride

Supplier: BeanTown Chemical

Synonyms: Chloroiodide, Iodochloride, Iodine chloride

141955-100G 141955-25G 141955-500G
BT141955-100GEA 87.2 USD
BT141955-100G BT141955-25G BT141955-500G
Iodine monochloride
Iodine monochloride

CAS: 7790-99-0; EC No: 232-236-7; MDL No: MFCD00011354; RTECS: NN1650000 UN No: UN3498; Haz Class: 8; Packing Group: II Crystalline/Liquid; Linear Formula: ICl; MW: 162.36 Boiling Point: 96-98° Density (g/mL): 3.24 Light Sensitive, Moisture Sensitive


  • RTECS: NN1650000
  • Light Sensitive, Moisture Sensitive
  • Crystalline/Liquid

Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 97.3
Melting Pt: 26
Density: 3.24 g/mL
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 3498
ADR: 8 II
Merck Index: 13,05039

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR