Dibutyltin dichloride ≥96%

Supplier: BeanTown Chemical

Synonyms: Dichlorodibutylstannane, Di-n-butyltin dichloride, DBTC

141775-10G 141775-250G 141775-50G
BT141775-10GEA 22 USD
BT141775-10G BT141775-250G BT141775-50G
Dibutyltin dichloride ≥96%
Dibutyltin dichloride

CAS: 683-18-1; EC No: 211-670-0; MDL No: MFCD00000518; RTECS: WH7100000 UN No: UN2928; Haz Class: 6.1 (8); Packing Group: II Crystalline; Linear Formula: (CH3CH2CH2CH2)2SnCl2; Molecular Formula: C8H18Cl2Sn; MW: 303.83 Melting Point: 37-40°; Boiling Point: 135°/10 mmHg; Flash point: 110°C (230°F) Moisture Sensitive


  • RTECS: WH7100000
  • Moisture Sensitive
  • Crystalline

Formula: C₈H₁₈Cl₂Sn
MW: 303.85 g/mol
Boiling Pt: 275
Melting Pt: 37…40 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000518
CAS Number: 683-18-1
EINECS: 211-670-0
UN: 2928
ADR: 6.1 II

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR