Formaldehyde 37% w/w water

Supplier: BeanTown Chemical

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

141265-100ML 141265-2.5L 141265-500ML
BT141265-100MLEA 27.71 USD
BT141265-100ML BT141265-2.5L BT141265-500ML
Formaldehyde 37% w/w water
Formaldehyde

CAS: 50-00-0; EC No: 200-001-8; MDL No: MFCD00003274; RTECS: LP8925000 UN No: UN1198; Haz Class: 3 (8); Packing Group: III Liquid; Linear Formula: HCHO; MW: 30.03 Flash point: 64°C (147°F) Density (g/mL): 1.09; Refractive Index: 1.376


  • RTECS: LP8925000
  • Refractive index: 1.376
  • Liquid

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Density: 1.09 g/mL
Flash Pt: 64 °C (147 °F)
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 1198
ADR: 8 III
Merck Index: 13,04259

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR