Molybdenum(VI) oxide ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Molybdenum trioxide
140765-100G
140765-25G
BT140765-100GEA
212.87
USD
BT140765-100G
BT140765-25G
Molybdenum(VI) oxide ≥99.995%
Molybdenum (VI) oxide
CAS: 1313-27-5; EC No: 215-204-7; MDL No: MFCD00003469; RTECS: QA4725000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: MoO3; MW: 143.94
Melting Point: 795°; Boiling Point: 1155° (sublimes)
Density (g/mL): 4.692
- RTECS: QA4725000
- Powder
Formula:
MoO₃ MW: 143.94 g/mol Boiling Pt: 1155 (subl.) Melting Pt: 795 Density: 4.692 g/mL |
MDL Number:
MFCD00003469 CAS Number: 1313-27-5 EINECS: 215-204-7 UN: 3288 ADR: 6.1 III Merck Index: 13,06261 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...