Oxalic acid 10% w/v water

Supplier: BeanTown Chemical
Danger

Synonyms: Ethanedioic acid

140555-1L 140555-4L
BT140555-1LEA 64.81 USD
BT140555-1L BT140555-4L
Oxalic acid 10% w/v water
Oxalic acid

CAS: 144-62-7; EC No: 205-634-3; MDL No: MFCD00002573 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Linear Formula: HOOCCOOH; Molecular Formula: C2H2O4; MW: 90.04


  • Liquid

Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3265
ADR: 8 III
Merck Index: 13,06980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR