Methylenediphosphonic acid ≥99%

Supplier: BeanTown Chemical

140525-1G 140525-25G 140525-5G
BT140525-1GEA 83.98 USD
BT140525-1G BT140525-25G BT140525-5G
Methylenediphosphonic acid ≥99%
Methylenediphosphonic acid

CAS: 1984-15-2; EC No: 217-851-0; MDL No: MFCD00002139; RTECS: SZ9138000 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: CH2[P(O)(OH)2]2; Molecular Formula: CH6O6P2; MW: 176.00 Melting Point: 197-199°


  • RTECS: SZ9138000
  • Crystalline

Formula: CH₆O₆P₂
Melting Pt: 197…199 °C
MDL Number: MFCD00002139
CAS Number: 1984-15-2
EINECS: 217-851-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR