Ammonium perrhenate ≥99.999%
Supplier: BeanTown Chemical
140395-1G
140395-5G
BT140395-1GEA
55
USD
BT140395-1G
BT140395-5G
Ammonium perrhenate ≥99.999%
Ammonium perrhenate
CAS: 13598-65-7; EC No: 237-075-6; MDL No: MFCD00012377; RTECS: VI0710000
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: NH4ReO4; MW: 268.24
Density (g/mL): 3.97
- RTECS: VI0710000
- Crystalline
Formula:
H₄NO₄Re MW: 268.24 g/mol Density: 3.97 g/mL |
MDL Number:
MFCD00012377 CAS Number: 13598-65-7 EINECS: 237-075-6 UN: 1479 ADR: 5.1 II |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...