Calcium oxide ≥99.995% (metals basis) (excluding other alkaline earth and alkali metals)
Supplier: BeanTown Chemical
Synonyms:
Quicklime, Calcium oxide from marble, caustic, Lime
139885-1G
139885-25G
139885-5G
BT139885-1GEA
19
USD
BT139885-1G
BT139885-25G
BT139885-5G
Calcium oxide ≥99.995% (metals basis) (excluding other alkaline earth and alkali metals)
Calcium oxide
CAS: 1305-78-8; EC No: 215-138-9; MDL No: MFCD00010911; RTECS: EW3100000
UN No: UN1910; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: CaO; MW: 56.08
Boiling Point: 2850°
Density (g/mL): 3.3
Air Sensitive, Moisture Sensitive
- RTECS: EW3100000
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
CaO Boiling Pt: 2850 °C Density: 3.3 g/mL |
MDL Number:
MFCD00010911 CAS Number: 1305-78-8 EINECS: 215-138-9 ADR: 8,III |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...