Trifluoroacetic acid ≥99.5%
Supplier: BeanTown Chemical
|
Danger
|
Synonyms:
TFA
139770-100G
139770-2.5KG
139770-500G
BT139770-100GEA
71.86
USD
BT139770-100G
BT139770-2.5KG
BT139770-500G
Trifluoroacetic acid ≥99.5%
Trifluoroacetic acid
CAS: 76-05-1; EC No: 200-929-3; MDL No: MFCD00004169; RTECS: AJ9625000
UN No: UN2699; Haz Class: 8; Packing Group: I
Liquid; Linear Formula: CF3COOH; MW: 114.02
Melting Point: -15°; Boiling Point: 72-73°
Density (g/mL): 1.489; Refractive Index: 1.284
Hygroscopic
- RTECS: AJ9625000
- Refractive index: 1.284
- Hygroscopic
Formula:
C₂HF₃O₂ MW: 114.02 g/mol Boiling Pt: 71.8 Melting Pt: -15 Density: 1.489 |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8 I Merck Index: 14,9681 |
|
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...