N-Ethylmaleimide (NEM) ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione
139675-1G
139675-25G
139675-5G
BT139675-1GEA
23
USD
BT139675-1G
BT139675-25G
BT139675-5G
N-Ethylmaleimide (NEM) ≥99%
N-Ethylmaleimide
CAS: 128-53-0; EC No: 204-892-4; MDL No: MFCD00005509; RTECS: UX9625000
UN No: UN2928; Haz Class: 6.1 (8); Packing Group: II
Crystalline; Molecular Formula: C6H7NO2; MW: 125.13
Melting Point: 43-46°; Boiling Point: 209-211°; Flash point: 73°C (163°F)
Light Sensitive
- RTECS: UX9625000
- Light Sensitive
- Crystalline
Formula:
C₆H₇NO₂ Boiling Pt: 209…211 °C Melting Pt: 43…46 °C Flash Pt: 73 °C (163 °F) |
MDL Number:
MFCD00005509 CAS Number: 128-53-0 EINECS: 204-892-4 UN: 2928 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...