n-Hexane, anhydrous

Supplier: BeanTown Chemical

Synonyms: Hexyl hydride, Hexanen, Hexane

139290-1L 139290-250ML 139290-4L
BT139290-1LEA 87.96 USD
BT139290-1L BT139290-250ML BT139290-4L
n-Hexane, anhydrous
n-Hexane

CAS: 110-54-3; EC No: 203-777-6; MDL No: MFCD00009520; RTECS: MN9275000 UN No: UN1208; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3(CH2)4CH3; Molecular Formula: C6H14; MW: 86.18 Melting Point: -95°; Boiling Point: 69°; Flash point: -26°C (-15°F) Density (g/mL): 0.659; Refractive Index: 1.375


  • RTECS: MN9275000
  • Refractive index: 1.375
  • Liquid

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/mL
Flash Pt: –26 °C (–15 °F)
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR