Toluene ≥99.5% ACS
Supplier: BeanTown Chemical
CAS: 108-88-3; EC No: 203-625-9; MDL No: MFCD00008512; RTECS: XS5250000
UN No: UN1294; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: C6H5CH3; Molecular Formula: C7H8; MW: 92.14
Melting Point: -93°; Boiling Point: 110-111°; Flash point: 4°C (39°F)
Density (g/mL): 0.865; Refractive Index: 1.496
- RTECS: XS5250000
- Refractive index: 1.496
- Liquid
Formula:
C₆H₅CH₃ Boiling Pt: 110…111 °C Melting Pt: –93 °C Density: 0.865 g/mL Flash Pt: 4 °C (39 °F) |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II |
|
Specification Test Results
Assay | 99.5% min. |
Color (APHA) | ≤10ppm |
Residue after evaporation | ≤10ppm |
Substances darkened by H2SO4 | Passes test |
Refractive index at 25°C | Passes test |
Sulfur compounds (as S) | ≤0.003% |
Water | ≤0.03% |
Benzene | ≤0.03% |
Total C8 aromatics | Per lot |
Non-aromatics | Per lot |
Specific gravity 60/60F | 0.869-0.873 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...