Ethyl (methylthio)acetate ≥98%

Supplier: BeanTown Chemical

Synonyms: (Methylthio)acetic ethyl ester, Ethyl 2-(methylthio)acetate

138520-100G 138520-25G 138520-5G
BT138520-100GEA 145 USD
BT138520-100G BT138520-25G BT138520-5G
Ethyl (methylthio)acetate ≥98%
Ethyl (methylthio)acetate

CAS: 4455-13-4; EC No: 224-700-2; MDL No: MFCD00009182 UN No: UN3272; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3SCH2CO2CH2CH3; Molecular Formula: C5H10O2S; MW: 134.20 Boiling Point: 70-72°/25 mmHg; Flash point: 59°C (138°F) Density (g/mL): 1.043; Refractive Index: 1.459 Moisture Sensitive


  • Refractive index: 1.459
  • Moisture Sensitive
  • Liquid

Formula: C₅H₁₀O₂S
MW: 134.2
Boiling Pt: 70
Density: 1.043 g/mL
Flash Pt: 59 °C (138 °F)
MDL Number: MFCD00009182
CAS Number: 4455-13-4
EINECS: 224-700-2
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR