Chlorotrimethylsilane ≥98%
Supplier: BeanTown Chemical
Synonyms:
TMCS, TMSC, Trimethylchlorosilane
138515-100ML
138515-25ML
138515-500ML
BT138515-100MLEA
31.45
USD
BT138515-100ML
BT138515-25ML
BT138515-500ML
Chlorotrimethylsilane ≥98%
Chlorotrimethylsilane
CAS: 75-77-4; EC No: 200-900-5; MDL No: MFCD00000502; RTECS: VV2710000
UN No: UN1298; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: (CH3)3SiCl; Molecular Formula: C3H9ClSi; MW: 108.64
Melting Point: -40°; Boiling Point: 57°; Flash point: -28°C (-18°F)
Density (g/mL): 0.856; Refractive Index: 1.387
Moisture Sensitive
- RTECS: VV2710000
- Refractive index: 1.387
- Moisture Sensitive
- Liquid
Formula:
C₃H₉ClSi Boiling Pt: 57 °C Melting Pt: –40 °C Density: 0.856 g/mL Flash Pt: –28 °C (–18 °F) |
MDL Number:
MFCD00000502 CAS Number: 75-77-4 EINECS: 200-900-5 UN: 1298 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...