Platinum(IV) chloride ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Platinum tetrachloride
138430-1G
138430-5G
BT138430-1GEA
158.63
USD
BT138430-1G
BT138430-5G
Platinum(IV) chloride ≥99.9%
Platinum (IV) chloride
CAS: 13454-96-1; EC No: 236-645-1; MDL No: MFCD00011182
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline/Powder; Linear Formula: PtCl4; MW: 336.89
Melting Point: 370° (decomposes)
Density (g/mL): 4.303
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
Cl₄Pt MW: 336.89 g/mol Melting Pt: 370 (dec.) Density: 4.303 g/mL |
MDL Number:
MFCD00011182 CAS Number: 13454-96-1 EINECS: 236-645-1 UN: 3260 ADR: 8 III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...