Potassium hydroxide 50% w/v water

Supplier: BeanTown Chemical

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

138400-1L 138400-4L 138400-500ML
BT138400-1LEA 80.25 USD
BT138400-1L BT138400-4L BT138400-500ML
Potassium hydroxide 50% w/v water
Potassium hydroxide

CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553; RTECS: TT2100000 UN No: UN1814; Haz Class: 8; Packing Group: II Liquid; Linear Formula: KOH; MW: 56.11 Density (g/mL): 1.456


  • RTECS: TT2100000
  • Liquid

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.456 g/mL
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR