Potassium-tert-butoxide ≥97%

Supplier: BeanTown Chemical

Synonyms: Potassium tert-butanolatePotassium t-butoxidePotassium tert-butoxidePotassium tert-butylate

138320-100G 138320-250G 138320-25G
BT138320-250GEA 72.88 USD
BT138320-250G BT138320-25G BT138320-100G
Potassium-tert-butoxide ≥97%
Potassium-tert-butoxide

  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: C₄H₉KO
MW: 112.21 g/mol
Melting Pt: 256…258 °C (dec.)
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR