Platinum, Plasma Standard Solution, Pt 1000 µg/ml

Supplier: BeanTown Chemical
138080-50ML 138080-100ML 138080-500ML
BT138080-50MLEA 138.6 USD
BT138080-50ML BT138080-500ML BT138080-100ML
Platinum, Plasma Standard Solution, Pt 1000 µg/ml
Standards Cationic Standards Platinum Standards
EC No: 231-595-7
Haz class: 8
Packing group: II
GHS symbol: GHS05
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR