Sodium selenite, anhydrous ≥99%

Supplier: BeanTown Chemical

137760-100G 137760-500G
BT137760-100GEA 80.23 USD
BT137760-100G BT137760-500G
Sodium selenite, anhydrous ≥99%
Sodium selenite

CAS: 10102-18-8; EC No: 233-267-9; MDL No: MFCD00003489; RTECS: VS7350000 UN No: UN2630; Haz Class: 6.1; Packing Group: I Crystalline/Powder; Linear Formula: Na2SeO3; MW: 172.94 Melting Point: 350° Hygroscopic


  • RTECS: VS7350000
  • Hygroscopic
  • Crystalline/Powder

Formula: Na₂SeO₃
MW: 172.94 g/mol
Melting Pt: 710
MDL Number: MFCD00003489
CAS Number: 10102-18-8
EINECS: 233-267-9
UN: 2630
ADR: 6.1 I
Merck Index: 12,08822

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR