n-Hexane ≥95.0% for HPLC

Supplier: BeanTown Chemical

Synonyms: Hexyl hydride, Hexanen, Hexane

137585-1L 137585-4L 137585-4X1L
BT137585-1LEA 69.4 USD
BT137585-1L BT137585-4L BT137585-4X1L
n-Hexane ≥95.0% for HPLC
n-Hexane

CAS: 110-54-3; EC No: 203-777-6; MDL No: MFCD00009520; RTECS: MN9275000 UN No: UN1208; Haz Class: 3; Packing Group: II Liquid, distilled in glass; Linear Formula: CH3(CH2)4CH3; Molecular Formula: C6H14; MW: 86.18 Melting Point: -95°; Boiling Point: 69°; Flash point: -26°C (-15°F) Density (g/mL): 0.659; Refractive Index: 1.375


  • RTECS: MN9275000
  • Refractive index: 1.375
  • Liquid, distilled in glass

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/mL
Flash Pt: –26 °C (–15 °F)
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 95.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR