Silver tetrafluoroborate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Argentous tetrafluoroborate
137490-10G
137490-50G
BT137490-10GEA
125.97
USD
BT137490-10G
BT137490-50G
Silver tetrafluoroborate ≥99%
Silver tetrafluoroborate
CAS: 14104-20-2; EC No: 237-956-5; MDL No: MFCD00003408; RTECS: ED2875000
UN No: UN3260; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: AgBF4; MW: 194.67
Melting Point: 70-73°
Light Sensitive, Moisture Sensitive
- RTECS: ED2875000
- Light Sensitive, Moisture Sensitive
- Crystalline
Formula:
AgBF₄ Melting Pt: 70…73 °C |
MDL Number:
MFCD00003408 CAS Number: 14104-20-2 EINECS: 237-956-5 UN: 3260 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...