Mercury(II) acetate ≥98%

Supplier: BeanTown Chemical

137345-100G 137345-25G 137345-500G
BT137345-100GEA 80.62 USD
BT137345-100G BT137345-25G BT137345-500G
Mercury(II) acetate ≥98%
Mercury (II) acetate

CAS: 1600-27-7; EC No: 216-491-1; MDL No: MFCD00012165; RTECS: AI8575000 UN No: UN1629; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: Hg(OOCCH3)2; MW: 318.68 Melting Point: 179-182° Density (g/mL): 3.28 Light Sensitive


  • RTECS: AI8575000
  • Light Sensitive
  • Powder

Formula: (H₃CCOO)₂Hg
Melting Pt: 179…182 °C
Density: 3.28 g/mL
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR