n-Hexane for spectrophotometry
Supplier: BeanTown Chemical
Synonyms:
Hexyl hydride, Hexanen, Hexane
137135-1L
137135-4L
137135-4X1L
BT137135-1LEA
46
USD
BT137135-1L
BT137135-4L
BT137135-4X1L
n-Hexane for spectrophotometry
n-Hexane
CAS: 110-54-3; EC No: 203-777-6; MDL No: MFCD00009520; RTECS: MN9275000
UN No: UN1208; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3(CH2)4CH3; Molecular Formula: C6H14; MW: 86.18
Melting Point: -95°; Boiling Point: 69°; Flash point: -26°C (-15°F)
Density (g/mL): 0.659; Refractive Index: 1.375
- RTECS: MN9275000
- Refractive index: 1.375
- Liquid
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 69 Melting Pt: -94.3 Density: 0.659 g/mL Flash Pt: –26 °C (–15 °F) |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3 II Merck Index: 13,04712 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...