2-Amino-1,3,4-thiadiazole ≥98%

Supplier: BeanTown Chemical

Synonyms: [1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine

137010-25G 137010-5G
BT137010-25GEA 162 USD
BT137010-25G BT137010-5G
2-Amino-1,3,4-thiadiazole ≥98%
2-Amino-1,3,4-thiadiazole

CAS: 4005-51-0; EC No: 223-657-7; MDL No: MFCD00003107; RTECS: XI3000000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C2H3N3S; MW: 101.13 Melting Point: 188-191° (decomposes)


  • RTECS: XI3000000
  • Powder

Formula: C₂H₃N₃S
MW: 101.13
Melting Pt: 188…191 °C (dec.)
MDL Number: MFCD00003107
CAS Number: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR