Yttrium ≥99.9% (trace rare earth metals basis), Powder -40 mesh
Supplier: BeanTown Chemical
CAS: 7440-65-5; EC No: 231-174-8; MDL No: MFCD00011468
UN No: UN3089; Haz Class: 4.1; Packing Group: II
Powder, packaged under argon; Molecular Formula: Y; MW: 88.91
Melting Point: 1522°; Boiling Point: 3338°
Density (g/mL): 4.469
- Powder, packaged under argon
Formula:
Y Boiling Pt: 3338 °C Melting Pt: 1522 °C Density: 4.469 g/mL |
MDL Number:
MFCD00011468 CAS Number: 7440-65-5 EINECS: 231-174-8 UN: 3089 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...