1-Chlorobutane ≥99.5% for HPLC

Supplier: BeanTown Chemical

Synonyms: n-Butyl Chloride

134620-4L 134620-250ML 134620-1L
BT134620-1LEA 85.34 USD
BT134620-1L BT134620-250ML BT134620-4L
1-Chlorobutane ≥99.5% for HPLC
1-Chlorobutane

  • RTECS: EJ6300000
  • Refractive index: 1.402
  • Liquid

Formula: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Boiling Pt: 77…78 °C
Melting Pt: –123 °C
Density: 0.886 g/mL
Flash Pt: –12 °C (10 °F)
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/58/18/725818.jpg

Order Now

Specification Test Results

Assay 99.5% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR