Copper(II) chloride dihydrate 99,0 ACS

Supplier: BeanTown Chemical

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

134590-100G 134590-2KG 134590-500G
BT134590-100GEA 55.62 USD
BT134590-100G BT134590-2KG BT134590-500G
Copper(II) chloride dihydrate 99,0 ACS
Copper (II) chloride dihydrate

CAS: 10125-13-0; EC No: 231-210-2; MDL No: MFCD00149792 UN No: UN2802; Haz Class: 8; Packing Group: III Granular; Linear Formula: CuCl2·2H2O; MW: 170.48 Melting Point: 100° (decomposes) Air Sensitive , Hygroscopic


  • Air Sensitive, Hygroscopic
  • Granular

Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 (dec.)
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8 III
Merck Index: 13,02660

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR