Hydrofluoric acid 48 - 51% ACS
Supplier: BeanTown Chemical
134430-100G
134430-500G
BT134430-100GEA
43.82
USD
BT134430-100G
BT134430-500G
Hydrofluoric acid 48 - 51% ACS
Hydrofluoric acid
CAS: 7664-39-3; EC No: 231-634-8; MDL No: MFCD00011346; RTECS: MW7875000
UN No: UN1790; Haz Class: 8 (6.1); Packing Group: II
Liquid; Molecular Formula: HF; MW: 20.01
Density (g/mL): 1.15
Hygroscopic
- RTECS: MW7875000
- Hygroscopic
- Liquid
Formula:
HF Density: 1.15 g/mL |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 EINECS: 231-634-8 UN: 1790 ADR: 8,II |
Specification Test Results
Assay | 48-51% |
Fluosilicic acid | ≤0.01% |
Residue after ignition | ≤5ppm |
Chloride (Cl) | ≤5ppm |
Phosphate (PO4) | ≤1ppm |
Sulfate & sulfite | ≤5ppm |
Arsenic (As) | ≤0.05ppm |
Copper (Cu) | ≤0.1ppm |
Heavy metals (as Pb) | ≤0.5ppm |
Iron (Fe) | ≤1ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...