Molybdenum(VI) oxide ≥99.9995%

Supplier: BeanTown Chemical

Synonyms: Molybdenum trioxide

134185-100G 134185-25G 134185-500G
BT134185-100GEA 282 USD
BT134185-100G BT134185-25G BT134185-500G
Molybdenum(VI) oxide ≥99.9995%
Molybdenum (VI) oxide

CAS: 1313-27-5; EC No: 215-204-7; MDL No: MFCD00003469; RTECS: QA4725000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: MoO3; MW: 143.94 Melting Point: 795°; Boiling Point: 1155° (sublimes) Density (g/mL): 4.692


  • RTECS: QA4725000
  • Powder

Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1155 (subl.)
Melting Pt: 795
Density: 4.692 g/mL
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1 III
Merck Index: 13,06261

Order Now

Specification Test Results

Assay 99.9995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR